MilliporeSigma
Synthesis of the oxygenated pactamycin core.
Organic letters (2007-03-07)
Spencer Knapp, Younong Yu
ABSTRACT
[structure: see text]. Pactamycin, one of the most complex and densely functionalized aminocyclitol antibiotics known, presents synthetic challenges that include reactivity and sterics, relative and absolute stereochemistry, and functional group compatibility and protection. An approach is reported that features four different types of (cyclopentane) face selective functionalization reactions and results in a polyfunctionalized and appropriately protected intermediate that incorporates all the core carbons and the oxygenated functionality of the target.