MilliporeSigma
Chemical changes in rubber allergens during vulcanization.
Contact dermatitis (2007-08-08)
Ola Bergendorff, Christina Persson, Anna Lüdtke, Christer Hansson
ABSTRACT
Allergic contact dermatitis to rubber is caused by residues of chemicals used in manufacturing a rubber product. Several different additives are used to achieve a final product of the desired characteristics. Accelerators such as thiurams, dithiocarbamates, and mercaptobenzothiazoles are often among the additives responsible for allergic reactions recognized by dermatologists. The chemistry of the vulcanization process is complicated; as it occurs at an elevated temperature with a mixture of reactive chemicals, the compositions of the initial and final products differ. This paper investigates the changes in composition of common allergens during vulcanization, doing so by chemically analysing various rubber formulations at different stages of the process. Major changes were found in which added chemicals were consumed and new ones produced. An important observation is that thiuram disulfides rarely appear in the final rubber although they may have been used as additives. Instead, thiurams are often converted to dithiocarbamates or to products formed by addition to mercaptobenzothiazole structures, if these have been used together with thiurams as accelerators.
Sigma-Aldrich
2,2′-Dithiobis(benzothiazole), 99%