MilliporeSigma
Preparation of diazabicyclo[4.3.0]nonene-based peptidomimetics.
The Journal of organic chemistry (2007-08-10)
Craig A Hutton, Paul A Bartlett
ABSTRACT
Several functionalized diazabicyclo[4.3.0]nonenes and other heterocycles have been prepared as potential peptidomimetic scaffolds. A novel and efficient method has been developed for the preparation of N-substituted gamma-lactams 13. Preparation of amidine-containing 1,5-diazabicyclo[4.3.0]nonenes 43 and 44 has been achieved through Hg-mediated cyclization of the precursor N-aminopropyl-gamma-thiolactams and subsequent functional group manipulation. Bicycle 43 represents a novel scaffold for potential peptide turn mimetics, whereas 44 could potentially be employed as an alpha-helix template attached to the C-terminus of peptides. These compounds are novel additions to the current range of small-molecule constrained peptidomimetics.
Sigma-Aldrich
1,5-Diazabicyclo[4.3.0]non-5-ene, purum, ≥98.0% (GC)
Sigma-Aldrich
1,5-Diazabicyclo[4.3.0]non-5-ene, 98%