MilliporeSigma
  • The effect of a metaphylactic treatment with diclazuril (Vecoxan) on the oocyst excretion and growth performance of calves exposed to a natural Eimeria infection.
The effect of a metaphylactic treatment with diclazuril (Vecoxan) on the oocyst excretion and growth performance of calves exposed to a natural Eimeria infection.
Veterinary parasitology (2007-09-14)
A Daugschies, J Agneessens, L Goossens, H Mengel, P Veys
ABSTRACT
A multicenter field efficacy study was performed in six farms located in Belgium, France and Germany with a history of suspected coccidiosis outbreaks and the proven presence of Eimeria bovis and/or Eimeria zuernii. At each of these trial sites the calves were randomly allocated to two groups (T, treated; C, control) of similar size. In total, 231 calves were included in the study. Group T calves (n=116) were drenched with a single dose of diclazuril (Vecoxan, 1mg/kg body weight) close to the time of expected outbreak of coccidiosis (day 1 of the study period), group C calves (n=115) served as placebo-treated controls. Although E. bovis and/or E. zuernii were identified at all trial sites, clinical coccidiosis was only noted in 16% of the group C calves. At day 5 of the study period (4 days after treatment), faecal oocyst counts (opg, oocysts per gram) were substantially lower in group T ("short-term effect") compared to group C. At four trial sites, the respective values of groups T and C were significantly different (P=0.0132 to P=0.0001) in favour of group T. For the pooled data of all trial sites, this effect was highly significant (P<0.0001). The overall faecal oocyst counts from day 3 until day 21 of the study period ("Area Under the Curve") was significantly reduced in group T by 87.2-99.5% ("long-term effect") at five trial sites (P=0.0139 to P<0.0001). The pooled data revealed a highly significant effect of treatment on oocyst excretion over the observation period (P<0.0001). On five of the six trial sites, the average weight gain was higher in group T than in group C. On those trial sites, the average weight gain of group T calves exceeded that of the controls by 95-268g/day and by 2.0-6.0kg over the study period. This effect was statistically significant (P<0.01) at one trial site. Altogether the calves of group T gained on average 129g more weight daily than the controls (+2.7kg over the study period). For these pooled data, statistical analysis confirmed the positive effect of treatment of calves exposed to coccidiosis on growth performance (P=0.003). In conclusion, metaphylactic treatment with diclazuril efficiently controls coccidiosis in calves thus reducing environmental contamination with oocysts and preventing negative effects of natural exposure to coccidiosis on growth performance of calves.
Product Description
Supelco
Diclazuril, VETRANAL®, analytical standard
Diclazuril for system suitability, European Pharmacopoeia (EP) Reference Standard