MilliporeSigma
  • Effects of flavoxate hydrochloride on voltage-dependent Ba2+ currents in human detrusor myocytes at different experimental temperatures.
Effects of flavoxate hydrochloride on voltage-dependent Ba2+ currents in human detrusor myocytes at different experimental temperatures.
Naunyn-Schmiedeberg's archives of pharmacology (2007-10-03)
Toshihisa Tomoda, Hai-Lei Zhu, Kazuomi Iwasa, Manami Aishima, Atsushi Shibata, Narihito Seki, Seiji Naito, Noriyoshi Teramoto
ABSTRACT
The inhibitory effects of flavoxate hydrochloride (piperidinoethyl-3-methylflavone-8-carboxylate; hereafter referred as flavoxate) on voltage-dependent nifedipine-sensitive inward Ba2+ currents (I Ba) in human detrusor myocytes were investigated at different temperatures using conventional whole-cell patch-clamp techniques. When the bath-solution temperature was increased from 22 degrees C to 30 degrees C, I Ba peak amplitude was enhanced by approximately twice at several test potentials. Neither the I Ba threshold nor the membrane potentials for the I Ba maximum peak amplitude was affected by the temperature change. The concentration-response curves of flavoxate at both 30 degrees C (Ki = 5.1 microM) and 37 degrees C (Ki = 4.6 microM) were slightly shifted to the left in comparison with that at 22 degrees C (Ki = 10.3 microM). Similar results were also obtained in the presence of nifedipine (Ki = 14 nM at 22 degrees C vs. Ki = 2.5 nM at 30 degrees C and Ki = 2.1 nM at 37 degrees C). Altering the bath-solution temperature from 22 degrees C to 30 degrees C shifted the steady-state inactivation curve of I Ba at -90 mV to the left. At 30 degrees C, the steady-state inactivation curve of I Ba in the presence of flavoxate was also shifted to the left in comparison with that in the absence of flavoxate. Either 3-isobutyl-1-methylxanthine (IBMX) or theophylline, a phosphodiesterase inhibitor, caused little effects on I Ba, although cyclic nucleotides (dibutyryl cAMP and 8-Br-cGMP) inhibited I Ba. These results suggest that the inhibitory actions of flavoxate on I Ba in human detrusor myocytes were slightly changed at different experimental temperatures and that flavoxate directly blocked voltage-dependent L-type Ca2+ channels, not through the inhibition of phosphodiesterase activity pathway.