MilliporeSigma
  • Lithium overdose causing non-convulsive status epilepticus--the importance of lithium levels and the electroencephalography in diagnosis.
Lithium overdose causing non-convulsive status epilepticus--the importance of lithium levels and the electroencephalography in diagnosis.
Hong Kong medical journal = Xianggang yi xue za zhi (2007-12-07)
K K Yip, W T Yeung
ABSTRACT
We report a case of lithium overdose in a patient who presented in non-convulsive status epilepticus. The lithium toxicity was probably due to interaction with Moduretic. The diagnosis was not suspected until electroencephalography was performed. This case underscores the importance of therapeutic drug level monitoring of lithium, especially where toxicity is suspected, and the indispensable role electroencephalography plays by allowing a correct diagnosis to be made promptly.
Sigma-Aldrich
Lithium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Lithium sulfate, purum p.a., ≥98.0% (T)
Sigma-Aldrich
Lithium sulfate, ≥98.5% (titration)