MilliporeSigma
  • Maturation of Streptococcus pneumoniae lipoproteins by a type II signal peptidase is required for ABC transporter function and full virulence.
Maturation of Streptococcus pneumoniae lipoproteins by a type II signal peptidase is required for ABC transporter function and full virulence.
Molecular microbiology (2007-12-19)
Suneeta Khandavilli, Karen A Homer, Jose Yuste, Shilpa Basavanna, Timothy Mitchell, Jeremy S Brown
ABSTRACT
Cell surface lipoproteins are important for the full virulence of several bacterial pathogens, including Streptococcus pneumoniae. Processing of prolipoproteins seems to be conserved among different bacterial species, and requires type II signal peptidase (Lsp) mediated cleavage of the N-terminal signal peptide to form the mature lipoprotein. Lsp has been suggested as a target for new antibiotic therapies, but at present there are only limited data on the function of Lsp for Gram-positive bacterial pathogens. We have investigated the function and role during disease pathogenesis of the S. pneumoniae Lsp, which, blast searches suggest, is encoded by the gene Sp0928. Expression of Sp0928 protected Escherichia coli against the Lsp antagonist globomycin, and proteomics and immunoblot analysis demonstrated that deletion of Sp0928 prevented processing of S. pneumoniae prolipoproteins to mature lipoproteins. These data strongly suggest that Sp0928 encodes the S. pneumoniae Lsp. However, immunoblots of membrane-associated proteins, immunoelectron microscopy and flow cytometry assays all confirmed that in the absence of Lsp, immature lipoproteins were still attached to the cell surface. Despite preservation of lipoprotein attachment to the cell membrane, loss of S. pneumoniae Lsp resulted in several phenotypes associated with impaired lipoprotein function and reduced S. pneumoniae replication in animal models of infection.
Sigma-Aldrich
Globomycin from Streptomyces hagronensis