MilliporeSigma
  • Bacteriorhodopsin/amphipol complexes: structural and functional properties.
Bacteriorhodopsin/amphipol complexes: structural and functional properties.
Biophysical journal (2008-01-15)
Yann Gohon, Tassadite Dahmane, Rob W H Ruigrok, Peter Schuck, Delphine Charvolin, Fabrice Rappaport, Peter Timmins, Donald M Engelman, Christophe Tribet, Jean-Luc Popot, Christine Ebel
ABSTRACT
The membrane protein bacteriorhodopsin (BR) can be kept soluble in its native state for months in the absence of detergent by amphipol (APol) A8-35, an amphiphilic polymer. After an actinic flash, A8-35-complexed BR undergoes a complete photocycle, with kinetics intermediate between that in detergent solution and that in its native membrane. BR/APol complexes form well defined, globular particles comprising a monomer of BR, a complete set of purple membrane lipids, and, in a peripheral distribution, approximately 2 g APol/g BR, arranged in a compact layer. In the absence of free APol, BR/APol particles can autoassociate into small or large ordered fibrils.
Sigma-Aldrich
Octyl β-D-1-thioglucopyranoside, ≥98.0% (GC)