MilliporeSigma
Influence of kavain on hepatic ultrastructure.
World journal of gastroenterology (2008-01-19)
Shuang Fu, Emine Korkmaz, Filip Braet, Quan Ngo, Iqbal Ramzan
ABSTRACT
To investigate whether the major kavalactone kavain imposes adverse effects on the liver ultrastructure and function by affecting vascular and microvascular architecture and altering hepatocellular morphology. Kavain solution (10 microg/mL or 43.5 micromol/L) was perfused for 2 h in isolated rat livers. After standard fixation and tissue preparation, the samples were examined by scanning electron microscopy (SEM), transmission electron microscopy (TEM), and light microscopy (LM). LM, SEM, and TEM examinations indicated kavain-treated rat livers (n=4) displayed severe vascular and endothelial damage compared to control livers (n=4). The data so far support the hypothesis that kavain induces adverse effects on liver; additional investigations with other kavalactones and their effects on liver are urgently needed.