MilliporeSigma
  • Two new maltol glycosides and cyanogenic glycosides from Elsholtzia rugulosa Hemsl.
Two new maltol glycosides and cyanogenic glycosides from Elsholtzia rugulosa Hemsl.
Journal of natural medicines (2008-04-12)
Haizhou Li, Tatsuya Nakashima, Takashi Tanaka, Ying-Jun Zhang, Chong-Ren Yang, Isao Kouno
ABSTRACT
Two new maltol glycosides, maltol 6'-O-beta-D-apiofuranosyl-beta-D-glucopyranoside and maltol 6'-O-(5-O-p-coumaroyl)-beta-D-apiofuranosyl-beta-D-glucopyranoside, were isolated from Elsholtzia rugulosa Hemsl. along with 11 known compounds including prunasin and amygdalin. The structures were determined on the basis of spectroscopic and chemical evidence. This is the second example of isolation of cyanogenic glycosides from Lamiaceous plants.
Sigma-Aldrich
3-Hydroxy-2-methyl-4-pyrone, 99%
Sigma-Aldrich
Maltol, ≥99.0%, FCC, FG
Sigma-Aldrich
Maltol, natural, FG