MilliporeSigma
  • CADA, a potential anti-HIV microbicide that specifically targets the cellular CD4 receptor.
CADA, a potential anti-HIV microbicide that specifically targets the cellular CD4 receptor.
Current HIV research (2008-05-14)
Kurt Vermeire, Joachim Brouwers, Yven Van Herrewege, Roger Le Grand, Guido Vanham, Patrick Augustijns, Thomas W Bell, Dominique Schols
ABSTRACT
The cyclotriazadisulfonamide (CADA) compounds are a new class of specific CD4-targeted HIV entry inhibitors. The in vitro anti-HIV activity of CADA was shown to correlate with its ability to specifically downmodulate cell surface expression of the CD4 receptor in human cells. Here, we evaluated its potential as an anti-HIV microbicide. CADA exerted a clear CD4 receptor downregulating effect in dendritic cells (DC) and subsequently inhibited HIV-1(BaL) replication in DC/T cell co-cultures. The compound proved to be active against a variety of clinical isolates belonging to the HIV-1 subtypes A, B, C, D, F, G, H, AE and O. Furthermore, it prevented human T cells from being infected with the laboratory-adapted strains X4 HIV-1(NL4.3) and R5 SIV(mac251). Flow cytometric analysis demonstrated a significant and dose-dependent downregulation of CD4 on macaque PBMCs. In addition, the compound exerted a marked anti-SIV(mac251) activity in these cells from simian origin. The combination of CADA with cellulose acetate phthalate (CAP) resulted in a synergistic inhibition of HIV-1 and SIV infection. Finally, gel formulated CADA proved to preserve the CD4 downmodulating and antiviral activity of this compound when formulated as a microbicide gel. Thus, our data suggest that CADA may have potential as a broad-spectrum anti-HIV microbicide drug candidate. The preservation of the activity of gel formulated CADA will make it now feasible for testing this unique entry inhibitor in non-human primates, not only as a single drug but also in a synergistic conjunction with other anti-HIV compounds.
Product Description
Supelco
Cellulose acetate phthalate