MilliporeSigma
  • Sakuranetin induces adipogenesis of 3T3-L1 cells through enhanced expression of PPARgamma2.
Sakuranetin induces adipogenesis of 3T3-L1 cells through enhanced expression of PPARgamma2.
Biochemical and biophysical research communications (2008-06-05)
Takeshi Saito, Daigo Abe, Keizo Sekiya
ABSTRACT
Sakuranetin (5,4'-dihydroxy-7-methoxyflavone) belongs to the flavanone class of polyphenols predominantly known as phytoalexin in rice plant. In this study, we demonstrate that sakuranetin strongly induces differentiation of 3T3-L1 preadipocytes, as evidenced by increased triglyceride accumulation and glycerol-3-phosphate dehydrogenase (GPDH) activity. In addition, even in the absence of adipogenic hormonal stimuli, sakuranetin strongly induced adipogenesis and expression of genes that are critical for the adipocytes phenotype. Time-course analyses indicated that sakuranetin induces PPARgamma2 expression without prior induction of C/EBPbeta, a transcriptional regulator of PPARgamma2 in adipogenesis. In 3T3-L1 preadipocytes, the transcriptional factors GATA-2 and GATA-3 are known to down-regulate adipogenesis by direct binding to the C/EBPbeta protein and to the GATA-binding site on the PPARgamma2 promoter. We found that sakuranetin significantly reduced the expression of GATA-2. Moreover, we observed that sakuranetin stimulated glucose uptake in differentiated 3T3-L1 adipocytes. These results suggest that sakuranetin may contribute to maintain glucose homeostasis in animals.