MilliporeSigma
  • A direct and stereospecific approach to the synthesis of alpha-glycosyl thiols.
A direct and stereospecific approach to the synthesis of alpha-glycosyl thiols.
Organic & biomolecular chemistry (2008-06-06)
Ravindra T Dere, Yingxi Wang, Xiangming Zhu
ABSTRACT
A new simple method for the synthesis of alpha-glycosyl thiols is described. Ring-opening of 1,6-anhydrosugars with commercially available bis(trimethylsilyl)sulfide under the action of catalytic amounts of TMSOTf smoothly afforded alpha-glycosyl thiols in very high yields and in a stereospecific way.
Sigma-Aldrich
Trimethylsilyl trifluoromethanesulfonate, purum, ≥98.0% (T)
Sigma-Aldrich
Trimethylsilyl trifluoromethanesulfonate, 99%