MilliporeSigma
  • Gas chromatography/electron-capture negative ion mass spectrometry for the quantitative determination of 2- and 3-hydroxy fatty acids in bovine milk fat.
Gas chromatography/electron-capture negative ion mass spectrometry for the quantitative determination of 2- and 3-hydroxy fatty acids in bovine milk fat.
Journal of agricultural and food chemistry (2008-06-24)
Ramona Jenske, Walter Vetter
ABSTRACT
2- and 3-hydroxy fatty acids (2- and 3-OH-FAs) are bioactive substances reported in sphingolipids and bacteria. Little is known of their occurrence in food. For this reason, a method suitable for the determination of OH-FAs at trace levels in bovine milk fat was developed. OH-FAs (and conventional fatty acids in samples) were converted into methyl esters and the hydroxyl group was derivatized with pentafluorobenzoyl (PFBO) chloride to give PFBO- O-FA methyl esters. These derivatives with strong electron affinity were determined by gas chromatography interfaced to mass spectrometry using electron-capture negative ion in the selected ion monitoring mode (GC/ECNI-MS-SIM). This method proved to be highly sensitive and selective for PFBO-O-FA methyl esters. For the analysis of samples, two internal standards were used. For this purpose, 9,10-dideutero-2-OH-18:0 methyl ester (ISTD-1) from 2-OH-18:1(9 c) methyl ester as well as the ethyl ester of 3-PFBO-O-12:0 (ISTD-2) was synthesized. ISTD-1 served as a recovery standard whereas ISTD-2 was used for GC/MS measurements. The whole-sample cleanup consisted of accelerated solvent extraction of dry bovine milk, addition of ISTD 1, saponification, conversion of fatty acids into methyl esters by use of boron trifluoride, separation of the methyl esters of OH-FAs from nonsubstituted FAs on activated silica, conversion of OH-FAs methyl esters into PFBO-O-FA methyl esters, addition of ISTD-2, and measurement by GC/ECNI-MS-SIM. By this method, ten OH-FAs were quantified in bovine milk fat with high precision in the range from 0.02 +/- 0.00 to 4.49 +/- 0.29 mg/100 g of milk fat.
Sigma-Aldrich
2,3,4,5,6-Pentafluorobenzoyl chloride, 99%