MilliporeSigma
  • Characterization of the brain-specific non-AT(1), non-AT(2) angiotensin binding site in the mouse.
Characterization of the brain-specific non-AT(1), non-AT(2) angiotensin binding site in the mouse.
European journal of pharmacology (2008-06-24)
Vardan T Karamyan, Florian Gembardt, Felicia M Rabey, Thomas Walther, Robert C Speth
ABSTRACT
In the present study the existence of a non-AT(1), non-AT(2) angiotensin (Ang) binding site unmasked by the organomercurial protease inhibitor p-chloromercuribenzoate (PCMB) was demonstrated in mouse brain membranes, consistent with observations previously reported in the rat (Karamyan and Speth, 2007b). The pharmacological specificity of the non-AT(1), non-AT(2) angiotensin binding site was similar to the rat brain: Sar(1)-Ile(8)-Ang II > Ang III >or= Ang II > Ang I> p-aminophenylalanine(6) Ang II> CGP42112 > Ang IV > Ang 1-7 congruent with shorter angiotensin fragments. Neurotensin, bradykinin, and luteinizing hormone-releasing hormone showed K(i) values >10 microM, while substance P and VIP had K(i) values of approximately 2 microM. The non-AT(1), non-AT(2) angiotensin binding site was not present in adrenal, liver or kidney. Subcellular fractionation showed a higher density of [(125)I]Ang II binding in plasma membrane (P2) fractions of cerebral cortex and hypothalamus relative to debris (P1) fractions. The binding site is present in the brains of mice in which the AT(1a), AT(1b), AT(2), Mas, and neprilysin (EC 3.4.24.11, neutral endopeptidase) was knocked out confirming that the binding site is not a heretofore described angiotensin receptor or neprilysin. These observations confirm that this novel Ang binding site is distinct from classical AT(1), AT(2), AT(4) and Ang 1-7 receptors while retaining a high specificity for angiotensins that act on the known angiotensin receptors. Whether this binding site functions as a novel receptor for angiotensins or a specific angiotensinase with variable functionality at different redox states will require further study.