MilliporeSigma
  • Influence of a diene impurity on the molecular structure of phosphate-containing polymers with medical applications.
Influence of a diene impurity on the molecular structure of phosphate-containing polymers with medical applications.
Chemical communications (Cambridge, England) (2008-07-16)
Lisbeth Grøndahl, Shuko Suzuki, Edeline Wentrup-Byrne
ABSTRACT
We have demonstrated that the unacknowledged presence of almost 30% diene impurity in some commercial phosphate monomers had not only a significant effect on the molecular structure (topology) of a series of synthesized polymers but the instability of the ester functionalities during these polymerizations resulted in unexpectedly complex co-polymer chemistry.
Sigma-Aldrich
Phosphoric acid 2-hydroxyethyl methacrylate ester, contains 700-1000 ppm monomethyl ether hydroquinone, 90%