MilliporeSigma
  • Formation and decay of tetrazane derivatives--a Car-Parrinello molecular dynamics study.
Formation and decay of tetrazane derivatives--a Car-Parrinello molecular dynamics study.
Physical chemistry chemical physics : PCCP (2008-07-26)
Christel Nonnenberg, Irmgard Frank
ABSTRACT
The complications during flight 510 of the Ariane Project were ascribed to problems in the upper stage engine that employs the bipropellant monomethylhydrazine (MMH) and nitrogen tetroxide (NTO). This has led to the question what conditions or reactions possibly cause an uncontrolled behaviour in the combustion process of MMH/NTO. We use first-principles molecular dynamics to investigate the reactions of the hypergolic mixture in different chemical situations. It was possible to observe the ultrafast redox reaction between the reactants on the timescale of an unconstrained simulation. We show that electrostatic attraction is crucial for the understanding of this reaction. Besides a cold reaction preceding the ignition, a reaction path leading to the highly reactive compound dimethyltetrazane could be identified.