MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • [Chemical constituents of the pericarp of Trichosanthes rosthornii Harms].
[Chemical constituents of the pericarp of Trichosanthes rosthornii Harms].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (1991-02-01)
Z Chao, J Liu
ABSTRACT
Nine compounds were isolated from the pericarp of T. rosthornii and identified on the basis of spectral data to be palmitic acid, lignoceric acid, cerotic acid, montanic acid, melissic acid, L-(-)-alpha-monopalmitin, delta 7-stigmastenol, delta 7-stigmastenone-3 and delta 7-stigmatenol-3-beta-D-glucopyranoside. All but the last one are obtained from this species for the first time.
Sigma-Aldrich
DL-α-Palmitin, ≥99%