MilliporeSigma
  • Effect of borate buffer on the photolysis of riboflavin in aqueous solution.
Effect of borate buffer on the photolysis of riboflavin in aqueous solution.
Journal of photochemistry and photobiology. B, Biology (2008-09-02)
Iqbal Ahmad, Sofia Ahmed, Muhammad Ali Sheraz, Faiyaz H M Vaid
ABSTRACT
The photolysis of riboflavin (RF) in the presence of borate buffer (0.1-0.5M) at pH 8.0-10.5 has been studied using a specific multicomponent spectrophotometric method for the determination of RF and photoproducts, formylmethylflavin (FMF), lumichrome (LC) and lumiflavin (LF). The overall first-order rate constants for the photolysis of RF (1.55-4.36 x 10(-2)min(-1)) and the rate constants for the formation of FMF (1.16-3.52 x 10(-2)min(-1)) and LC (0.24-0.84 x 10(-2)min(-1)) have been determined. The values of all these rate constants decrease with an increase in buffer concentration suggesting the inhibition of photolysis reaction by borate species. The kinetic data support the formation of a RF-borate complex involving the ribityl side chain to cause the inhibition of photolysis. The second-order rate constants for the borate inhibited reaction range from 1.17-3.94 x 10(-2)M(-1)min(-1). The log k-pH profiles for the reaction at various buffer concentrations indicate a gradual increase in rate, with pH, up to 10 followed by a decrease in rate at pH 10.5 probably due to ionization of RF and quenching of fluorescence by borate species. A graph of second-order rate constants against pH is a sigmoid curve showing that the rate of photolysis increases with an increase in pH. The results suggest the involvement of excited singlet state, in addition to excited triplet state, in the formation of LC.
Sigma-Aldrich
Lumichrome