MilliporeSigma
  • Competitive binding of small ligands to nucleobases in AP site-containing DNA duplexes.
Competitive binding of small ligands to nucleobases in AP site-containing DNA duplexes.
Nucleic acids symposium series (2004) (2008-09-09)
Tomoe Kageyama, Yusuke Sato, Seiichi Nishizawa, Norio Teramae
ABSTRACT
By using lumichrome (Lch) as a masking ligand, we successfully control the binding selectivity of 2-amino-5,6,7-trimethyl-1,8-naphthyridine (ATMND) when binding to nucleobases in AP site-containing DNA duplexes (5'-TCT GCG TCC AGX GCA ACG CAC AC-3'/3'-AGA CGC AGG TCN CGT TGC GTG TG-5', X = AP site; Spacer C3, N = C or T). In solutions buffered to pH 7.0 (I = 0.11 M, at 5 degrees C), ATMND binds to cytosine and thymine with a comparable binding affinity (K(d) / nM: C: 7.7, T: 15). By contrast, in the presence of Lch, ATMND shows a clear binding selectivity for cytosine over thymine (K(d)/nM: C: 17, T: 204). Such competitive binding events are discussed with a view towards development of ligand-based fluorescence assay for single-nucleotide polymorphisms (SNPs) typing.
Sigma-Aldrich
Lumichrome