MilliporeSigma
  • Synthesis of a novel D-glucose-conjugated 15-crown-5 ether with a spiro ketal structure.
Synthesis of a novel D-glucose-conjugated 15-crown-5 ether with a spiro ketal structure.
Molecules (Basel, Switzerland) (2008-09-17)
Takashi Yamanoi, Yoshiki Oda, Hitomi Muraishi, Sho Matsuda
ABSTRACT
This paper describes a synthetic approach to a novel D-glucose-conjugated 15-crown-5 ether having a spiroketal structure starting from a 1-C-vinylated glucose derivative. The approach consists of the glycosylation of the vinylated glucose derivative to give an ethyleneoxy spacer derivative using bismuth(III) triflate, the conversion of the 1-C-vinyl group of the glucoside produced into a carboxylic acid group, and the intramolecular condensation between the carboxyl group and the terminal hydroxyl group in the ethyleneoxy spacer. A D-glucose-conjugated 15-crown-5 ether having a unique spiroketal structure was thus successfully synthesized.
Sigma-Aldrich
15-Crown-5, 98%