MilliporeSigma
  • Inconspicamide, new N-acylated serinol from the marine sponge Stelletta inconspicua.
Inconspicamide, new N-acylated serinol from the marine sponge Stelletta inconspicua.
Bioscience, biotechnology, and biochemistry (2008-11-11)
Reiko Ueoka, Tsuyoshi Fujita, Takashi Iwashita, Rob W M van Soest, Shigeki Matsunaga
ABSTRACT
A new N-acylated serinol, inconspicamide (1), was isolated from the marine sponge, Stelletta inconspicua, together with a glyceryl ether (2). Their structures were determined on the basis of spectroscopic data and the modified Mosher analysis. They exhibited moderate cytotoxic activity against HeLa human cervical cancer cells.
Sigma-Aldrich
Serinol, 98%