MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Effects of gabapentin and pregabalin on K+-evoked 3H-GABA and 3H-glutamate release from human neocortical synaptosomes.
Effects of gabapentin and pregabalin on K+-evoked 3H-GABA and 3H-glutamate release from human neocortical synaptosomes.
Naunyn-Schmiedeberg's archives of pharmacology (2008-11-13)
B Brawek, M Löffler, A Weyerbrock, T J Feuerstein
ABSTRACT
One site of action of the anticonvulsant, analgesic, and anxiolytic drugs gabapentin and pregabalin is the alpha(2)delta-subunit of voltage-sensitive Ca(2+) channels (VSCC). We therefore analyzed the effects of gabapentin and pregabalin on K(+)-evoked release of (3)H-gamma-aminobutyric acid (GABA) and (3)H-glutamate from superfused human neocortical synaptosomes. These neurotransmitters are released by Ca(2+)-dependent exocytosis and by Ca(2+)-independent uptake reversal. When a GABA transport inhibitor was present throughout superfusion to isolate exocytotic conditions, gabapentin and pregabalin (100 microM each) reduced K(+)-evoked (3)H-GABA release by 39% and 47%, respectively. These effects were antagonized by the alpha(2)delta-ligand L: -isoleucine (1 microM) suggesting the alpha(2)delta-subunit of terminal VSCC to mediate the reduction of exocytosis. Both drugs had no effect on exocytotic (3)H-glutamate release and also failed to modulate the release of (3)H-GABA and (3)H-glutamate caused by reversed uptake in the absence of external Ca(2+). Thus, an inhibition of glutamate release by gabapentin and pregabalin as main anticonvulsant principle is not supported by our experiments. An anticonvulsant mode of action of both drugs may be the reduction of a proconvulsant exocytotic GABA release.
Sigma-Aldrich
NO-711 hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
2-Amino-2-norbornanecarboxylic acid, amino acid transport inhibitor