MilliporeSigma
  • Vasorelaxant effects of forsythide isolated from the leaves of Forsythia viridissima on NE-induced aortal contraction.
Vasorelaxant effects of forsythide isolated from the leaves of Forsythia viridissima on NE-induced aortal contraction.
Phytomedicine : international journal of phytotherapy and phytopharmacology (2008-11-21)
T Iizuka, H Sakai, H Moriyama, N Suto, M Nagai, D Bagchi
ABSTRACT
Forsythide (F1) isolated from the leaves of Forsythia viridissima (Oleaceae) showed vasorelaxant effects on norepinephrine (NE)-induced contraction of rat aorta with or without endothelium. This compound did not affect contraction induced by high concentration potassium (60 mM K(+)) and phorbol 12,13-diacetate, but inhibited NE-induced contraction in the presence of nicardipine. These results demonstrated the inhibitory effects of F1 on NE-induced vasocontraction presumably due to decrease of calcium influx from extracellular area, which was induced by NE.