MilliporeSigma
  • A novel synthesis of highly substituted imidazo[1,5-a]pyrazine derivatives by 3-CR/2-CR sequence.
A novel synthesis of highly substituted imidazo[1,5-a]pyrazine derivatives by 3-CR/2-CR sequence.
Molecular diversity (2008-11-22)
Ahmad Shaabani, Ali Maleki, Hamid Mofakham
ABSTRACT
By combining a three-component reaction of 2, 3-diaminomaleonitrile, ketones and isocyanides with a subsequent reaction of 1,6-dihydropyrazine-2,3-dicarbonitrile derivatives obtained from isocyanide-based three-component reaction with various alkyl and aryl isocyanates or isothiocyanates can be assembled a new class of highly substituted imidazo[1,5-a]pyrazine derivatives.
Sigma-Aldrich
Diaminomaleonitrile, 98%