MilliporeSigma
  • Damage to the cytoplasmic membrane and cell death caused by dodine (dodecylguanidine monoacetate) in Pseudomonas syringae ATCC 12271.
Damage to the cytoplasmic membrane and cell death caused by dodine (dodecylguanidine monoacetate) in Pseudomonas syringae ATCC 12271.
Antimicrobial agents and chemotherapy (1991-02-01)
J P Cabral
ABSTRACT
Treatment of Pseudomonas syringae cells with low concentrations of the fungicide dodecylguanidine monoacetate (dodine) resulted in cell death and leakage of K+, UV-absorbing materials, and ribose-containing molecules. The results suggest that dodine causes gross and extensive damage to the cytoplasmic membrane, which is probably implicated in the death of cells.
Product Description
Supelco
Dodin, PESTANAL®, analytical standard