MilliporeSigma
  • 16,16-Dimethyl prostaglandin E2 efficacy on prevention and protection from bleomycin-induced lung injury and fibrosis.
16,16-Dimethyl prostaglandin E2 efficacy on prevention and protection from bleomycin-induced lung injury and fibrosis.
American journal of respiratory cell and molecular biology (2008-12-09)
Marco Failla, Tiziana Genovese, Emanuela Mazzon, Mary Fruciano, Evelina Fagone, Elisa Gili, Annalisa Barera, Cristina La Rosa, Enrico Conte, Nunzio Crimi, Salvatore Cuzzocrea, Carlo Vancheri
ABSTRACT
In this study, we evaluated the protective effect and therapeutic potential of the prostaglandin E(2) (PGE(2)) synthetic analog 16,16-dimethyl-PGE(2) (dmPGE(2)) in the animal model of pulmonary fibrosis induced by bleomycin. Mice subjected to intratracheal administration of bleomycin (1 mg/kg) received a dmPGE(2) dose of 30 microg/kg/day by continuous subcutaneous infusion. Bronchoalveolar lavage (BAL); immunohistochemical analysis for IL-1, TNF-alpha, and nitrotyrosine; measurement of fluid content in lung; myeloperoxidase activity assay; and lung histology were performed 1 week later. Lung histology and Sircol assay for collagen deposition were performed 3 weeks after treatments. Changes of body weight and survival rate were also evaluated at 1 and 3 weeks. Compared with bleomycin-treated mice, dmPGE(2) co-treated mice exhibited a reduced degree of body weight loss and mortality rate as well as of lung damage and inflammation, as shown by the significant reduction of: (1) lung infiltration by leukocytes; (2) myeloperoxidase activity; (3) IL-1, TNF-alpha, and nitrotyrosine immunostaining; (4) lung edema; and (5) histologic evidence of lung injury and collagen deposition. In a separate set of experiments, dmPGE(2) treatment was started 3 days after bleomycin administration, and the evaluation of lung damage and inflammation was assessed 4 days later. Importantly, delayed administration of dmPGE(2) also was able to protect from inflammation and lung injury induced by bleomycin. These results, indicating that dmPGE(2) is able to prevent and to reduce bleomycin-induced lung injury through its regulatory and anti-inflammatory properties, encourage further research to find new options for the treatment of pulmonary fibrosis.