MilliporeSigma
  • Successful pregnancy after gonadotropin-releasing hormone analogue and hysteroscopic myomectomy in a woman with diffuse uterine leiomyomatosis.
Successful pregnancy after gonadotropin-releasing hormone analogue and hysteroscopic myomectomy in a woman with diffuse uterine leiomyomatosis.
Archives of gynecology and obstetrics (2008-12-11)
Yoshihiko Shimizu, Hiroko Yomo, Nobuyuki Kita, Kentaro Takahashi
ABSTRACT
We report a patient with diffuse uterine leiomyomatosis, who wished to become pregnant. We performed hysteroscopic myomectomy after treatment with nafarelin acetate for 6 months. The patient conceived spontaneously soon after hysteroscopic myomectomy, and delivered a 2,798-g healthy baby.
Sigma-Aldrich
Nafarelin acetate salt hydrate, ≥95% (HPLC), solid