MilliporeSigma
  • DOCA and TGF-beta induce early growth response gene-1 (Egr-1) expression.
DOCA and TGF-beta induce early growth response gene-1 (Egr-1) expression.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2008-12-18)
Björn Friedrich, Andrea Janessa, Ferruh Artunc, Wilhelm Karl Aicher, Gerhard Anton Müller, Florian Lang, Teut Risler, Dorothea Alexander
ABSTRACT
Renal fibrosis is characterized by excessive accumulation of extracellular matrix proteins. Recent findings show that transforming growth factor-beta (TGF-beta) induces a rapid but transient expression of early growth response gene-1 (Egr-1) by skin fibroblasts. The present study aims to define the role of Egr-1 in mineralocorticoid-induced renal fibrosis. Therefore, we transiently transfected immortalized human renal fibroblasts (TK188) with recombinant Egr-1 and analysed the transcription of several pro-fibrotic genes (Coll1A1, Coll1A2, osteopontin, TIMP-1, and CTGF). We also examined Egr-1 expression and the regulation of pro-fibrotic genes in DOCA- (deoxycorticosterone acetate) and TGF-beta-treated renal fibroblasts. Finally, we compared Egr-1 gene expression in DOCA/high salt-induced fibrotic kidneys and untreated mice. Egr-1 transfection of TK188 fibroblasts induced the expression of TIMP-1 and osteopontin mRNA. Similar results were obtained after DOCA-activation of TK188 cells. Stimulation of TK188 with TGF-beta, but not with DOCA, resulted in elevated Coll1A1/Coll1A2 and CTGF levels. Co-stimulation with DOCA and TGF-beta was followed by enhanced Egr-1, Coll1A1, TIMP-1, and CTGF transcription. In conclusion, both DOCA and TGF-beta alone or in combination synergistically induced Egr-1 expression by human renal fibroblasts. DOCA induction of TIMP-1/osteopontin is Egr-1 dependent, whereas TGF-beta appears to induce Coll1A1 and CTGF by an Egr-1 independent pathway. In vivo analyses revealed significantly higher Egr-1 transcript levels in DOCA/high salt-induced fibrotic kidneys compared to untreated mice. Thus, we show for the first time that Egr-1 might participate in DOCA-induced renal fibrosis.
Product Description
Desoxycortone acetate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Deoxycorticosterone acetate