MilliporeSigma
  • Lactimidomycin, iso-migrastatin and related glutarimide-containing 12-membered macrolides are extremely potent inhibitors of cell migration.
Lactimidomycin, iso-migrastatin and related glutarimide-containing 12-membered macrolides are extremely potent inhibitors of cell migration.
Journal of the American Chemical Society (2009-01-10)
Jianhua Ju, Scott R Rajski, Si-Kyu Lim, Jeong-Woo Seo, Noël R Peters, F Michael Hoffmann, Ben Shen
ABSTRACT
Migrastatin (1), iso-migrastatin (5) and lactimidomycin (7) are all glutarimide-containing polyketides known for their unique structures and cytotoxic activities against human cancer cell lines. Migrastatin, a strong inhibitor of tumor cell migration, has been an important lead in the development of antimetastatic agents. Yet studies of the related 12-membered macrolides iso-migrastatin, lactimidomycin, and related analogues have been hampered by their limited availability. We report here the production, isolation, structural characterization, and biological activities of iso-migrastatin, lactimidomycin, and 23 related congeners. Our studies showed that, as a family, the glutarimide-containing 12-membered macrolides are extremely potent cell migration inhibitors with some members displaying activity on par or superior to that of migrastatin as exemplified by compounds 5, 7, and 9-12. On the basis of these findings, the structures and activity of this family of compounds as cell migration inhibitors are discussed.
Sigma-Aldrich
Glutarimide, 98%