MilliporeSigma
  • [Hexestrol inhibits ATP-induced contractile response of isolated preparations of pregnant human uterus].
[Hexestrol inhibits ATP-induced contractile response of isolated preparations of pregnant human uterus].
Eksperimental'naia i klinicheskaia farmakologiia (2009-01-15)
A U Ziganshin, D N Ablaeva, A A Khasanov, S I Ablaeva, D V Nurullina, L E Ziganshina
ABSTRACT
The results of in vitro experiments showed that hexestrol, a synthetic analog of estrogen hormones, at high concentrations inhibited the contractile response of isolated preparations of pregnant human uterus induced by ATP. It is suggested that estrogen hormones can interact with P2 receptors in human uterus during pregnancy.