MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • In vivo testing of nanoparticle-treated TTCP/DCPA-based ceramic surfaces.
In vivo testing of nanoparticle-treated TTCP/DCPA-based ceramic surfaces.
Acta biomaterialia (2009-01-16)
Wen-Cheng Chen, Chien-Ping Ju, Yin-Chun Tien, Jiin-Huey Chern Lin
ABSTRACT
This study reports the development of a non-dispersive calcium phosphate cement (nd-CPC) paste containing tetracalcium phosphate and anhydrous dicalcium phosphate that can be used as a filling material in dental and orthopedic applications. The nd-CPC bone cement is compared with two commercial materials, OsteoSet and Collagraft bone grafts. Gross examination of retrieved implants/bone composite samples indicated that none of the implants in this study evoked an inflammatory response. The OsteoSet (calcium sulfate) implant was resorbed too quickly to allow for osteo-remodeling, and it led to the formation of fibrous connective tissue in the fracture site, which remained even 24 weeks after implantation. Histological examination revealed that nd-CPC and Collagraft (hydroxyapatite/tricalcium phosphate/collagen) had greater remodeling and osteoconductive activity than OsteoSet at both 12 and 24 weeks after implantation. Greater remodeling activities were found with nd-CPC cement than with the other materials at 12 weeks after implantation, and the Fourier transform infrared absorption band of carbonate or cellulose derivatives grew from 6 weeks to 24 weeks after implantation in nd-CPC cement. These findings show that nd-CPC compares favorably to commercial bone remodeling materials, and the fact that it is in a paste formulation makes it an ideal material to fill regeneration defects.