MilliporeSigma
  • Photoirradiation products of flavin derivatives, and the effects of photooxidation on guanine.
Photoirradiation products of flavin derivatives, and the effects of photooxidation on guanine.
Bioorganic & medicinal chemistry letters (2009-03-04)
Katsuhito Kino, Teruhiko Kobayashi, Eiji Arima, Rie Komori, Takanobu Kobayashi, Hiroshi Miyazawa
ABSTRACT
Photoirradiation in the presence of riboflavin led to guanine oxidation and the formation of imidazolone. Meanwhile, riboflavin itself was degraded by ultraviolet light A (UV-A) and visible light (VIS) radiation, and the end product was lumichrome. VIS radiation in the presence of riboflavin oxidized guanine similarly to UV-A radiation. Although UV-A radiation with lumichrome oxidized guanine, VIS radiation with lumichrome did not. Thus, UV-A radiation with riboflavin can oxidize guanine even if riboflavin is degraded to lumichrome. In contrast, following VIS radiation degradation of riboflavin to lumichrome, VIS radiation with riboflavin is hardly capable of oxidizing guanine. The consequences of riboflavin degradation and guanine photooxidation can be extended to flavin mononucleotide and flavin adenine dinucleotide. In addition, we report advanced synthesis; carboxymethylflavin was obtained by oxidation of formylmethylflavin with chlorite and hydrogen peroxide; lumichrome was obtained by heating of formylmethylflavin in 50% AcOH; lumiflavin was obtained by incubation of formylmethylflavin in 2 M NaOH, followed by isolation by step-by-step concentration.
Sigma-Aldrich
Lumichrome