MilliporeSigma
  • Process simulation of single-step dimethyl ether production via biomass gasification.
Process simulation of single-step dimethyl ether production via biomass gasification.
Biotechnology advances (2009-04-28)
Fudong Ju, Hanping Chen, Xuejun Ding, Haiping Yang, Xianhua Wang, Shihong Zhang, Zhenghua Dai
ABSTRACT
In this study, we simulated the single-step process of dimethyl ether (DME) synthesis via biomass gasification using ASPEN Plus. The whole process comprised four parts: gasification, water gas shift reaction, gas purification, and single-step DME synthesis. We analyzed the influence of the oxygen/biomass and steam/biomass ratios on biomass gasification and synthesis performance. The syngas H(2)/CO ratio after water gas shift process was modulated to 1, and the syngas was then purified to remove H(2)S and CO(2), using the Rectisol process. Syngas still contained trace amounts of H(2)S and about 3% CO(2) after purification, which satisfied the synthesis demands. However, the high level of cold energy consumption was a problem during the purification process. The DME yield in this study was 0.37, assuming that the DME selectivity was 0.91 and that CO was totally converted. We performed environmental and economic analyses, and propose the development of a poly-generation process based on economic considerations.
Sigma-Aldrich
Dimethyl ether, ≥99%