- A novel mode of FoxM1 regulation: positive auto-regulatory loop.
A novel mode of FoxM1 regulation: positive auto-regulatory loop.
Cell cycle (Georgetown, Tex.) (2009-05-05)
Marianna Halasi, Andrei L Gartel
PMID19411834
ABSTRACT
Oncogenic transcription factor FoxM1 represents an attractive therapeutic target in the fight against cancer, because it is overexpressed in a majority of human tumors. Previously, we identified the thiazole antibiotics as potent inhibitors of FoxM1. Surprisingly, investigation of the mechanism of action of FoxM1 inhibitors revealed a novel mode of positive auto-regulation of FoxM1.