MilliporeSigma
  • Enantioselective addition of thioacetic acid to nitroalkenes via N-sulfinyl urea organocatalysis.
Enantioselective addition of thioacetic acid to nitroalkenes via N-sulfinyl urea organocatalysis.
Journal of the American Chemical Society (2009-06-06)
Kyle L Kimmel, MaryAnn T Robak, Jonathan A Ellman
ABSTRACT
The highly enantioselective addition of thioacetic acid to nitroalkenes using a new sulfinyl urea organocatalyst is described. The addition of thioacetic acid proceeds in high yields and enantioselectivities for a variety of aromatic and aliphatic nitroalkene substrates. This new method is useful for preparing chiral 1,2-aminothiol derivatives, as demonstrated by the first enantioselective synthesis of the clinically used antifungal drug sulconazole.
Sigma-Aldrich
Thioacetic acid, 96%
Sigma-Aldrich
Thioacetic acid, 96%
Sigma-Aldrich
Potassium thioacetate, 98%