MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Effect of corticotropin-like intermediate lobe peptide on presynaptic and postsynaptic glutamate receptors and postsynaptic GABA receptors in rat brain.
Effect of corticotropin-like intermediate lobe peptide on presynaptic and postsynaptic glutamate receptors and postsynaptic GABA receptors in rat brain.
Bulletin of experimental biology and medicine (2009-06-17)
V V Grigoriev, L N Petrova, T A Ivanova, A V Gabreliyan, T P Serkova
ABSTRACT
We studied the effect of corticotropin-like intermediate lobe peptide (CLIP) on presynaptic NMDA receptors and postsynaptic GABA, NMDA, and AMPA receptors in rat brain. CLIP inhibited presynaptic and postsynaptic NMDA receptors, but potentiated postsynaptic GABA and AMPA receptors. Our results indicate that CLIP modulates function of ionotropic receptors for glutamate and GABA.
Sigma-Aldrich
ProteoMass ACTH Fragment 18-39 MALDI-MS Standard, vial of 10 nmol, monoisotopic mol wt 2,464.1989 Da
Sigma-Aldrich
Adrenocorticotropic Hormone Fragment 18-39 human, ≥97% (HPLC)