MilliporeSigma
  • Gas-phase surface esterification of cellulose microfibrils and whiskers.
Gas-phase surface esterification of cellulose microfibrils and whiskers.
Biomacromolecules (2009-07-04)
Sophie Berlioz, Sonia Molina-Boisseau, Yoshiharu Nishiyama, Laurent Heux
ABSTRACT
A new and highly efficient synthetic method has been developed for the surface esterification of model cellulosic substrates of high crystallinity and accessibility, namely, freeze-dried tunicin whiskers and bacterial cellulose microfibrils dried by the critical point method. The reaction, which is based on the gas-phase action of palmitoyl chloride, was monitored by solid-state CP-MAS (13)C NMR. It was found that the grafting density not only depended on the experimental conditions, but also on the nature and conditioning of the cellulose samples. The structural and morphological modifications of the substrates at various degrees of grafting were revealed by scanning electron microscopy and X-ray diffraction analysis. These characterizations indicated that the esterification proceeded from the surface of the substrate to their crystalline core. Hence, for moderate degree of substitution, the surface was fully grafted whereas the cellulose core remained unmodified and the original fibrous morphology maintained. An almost total esterification could be achieved under certain conditions, leading to highly substituted cellulose esters, presenting characteristic X-ray diffraction patterns.
Sigma-Aldrich
Palmitoyl chloride, 98%