MilliporeSigma
  • Bioaugmentation of a sequencing batch reactor with Pseudomonas putida ONBA-17, and its impact on reactor bacterial communities.
Bioaugmentation of a sequencing batch reactor with Pseudomonas putida ONBA-17, and its impact on reactor bacterial communities.
Journal of hazardous materials (2009-07-07)
Fang-Bo Yu, Shinawar Waseem Ali, Li-Bo Guan, Shun-Peng Li, Shan Zhou
ABSTRACT
This study demonstrates the feasibility of using Pseudomonas putida ONBA-17 to bioaugment a sequencing batch reactor (SBR) treating o-nitrobenzaldehyde (ONBA) synthetic wastewater. To monitor its survival, the strain was chromosomally marked with gfp gene. After a transient adaptation, almost 100% degradation of ONBA was obtained within 8 days as compared with 23.47% of the non-inoculated control. The bioaugmented reactor has a better chemical oxygen demand (COD) removal performance (96.28%) than that (79.26%) of the control. The bioaugmentation not only enhanced the removal capability of target compound, but shortened system start-up time. After the increase in ONBA load, performance fluctuation of two reactors was observed, and the final treating effects of them were comparable. What is more, denaturing gradient gel electrophoresis (DGGE) analysis of 16S rRNA genes via a combination of pattern comparison and sequence phylogenetic analysis was performed to uncover changes in sludge microbial communities. Only the members of alpha, beta and gamma subdivisions of Proteobacteria were identified. To isolate ONBA-degrading relevant microorganisms, spread plate was used and four bacterial strains were obtained. Subsequent systematic studies on these bacteria characterized their traits which to some extent explained why such bacteria could be kept in the system. This study will help future research in better understanding of the bioreactor bioaugmentation.
Sigma-Aldrich
2-Nitrobenzaldehyde, 98%