MilliporeSigma
  • An efficient and economic asymmetric synthesis of (+)-nootkatone, tetrahydronootkatone, and derivatives.
An efficient and economic asymmetric synthesis of (+)-nootkatone, tetrahydronootkatone, and derivatives.
Organic letters (2009-07-28)
Anne M Sauer, William E Crowe, Gregg Henderson, Roger A Laine
ABSTRACT
A facile route to enantiomerically pure (+)-nootkatone and derivatives has been established through conjunctive stereoselective Grignard/anionic oxy-Cope (AOC) reactions.
Sigma-Aldrich
(+)-Nootkatone, ≥99.0% (GC)