MilliporeSigma
  • Stereoselective synthesis of spirooxindoles by palladium-catalyzed decarboxylative cyclization of gamma-methylidene-delta-valerolactones with isatins.
Stereoselective synthesis of spirooxindoles by palladium-catalyzed decarboxylative cyclization of gamma-methylidene-delta-valerolactones with isatins.
Organic letters (2009-07-30)
Ryo Shintani, Shin-ya Hayashi, Masataka Murakami, Momotaro Takeda, Tamio Hayashi
ABSTRACT
A new synthetic method of spirooxindole derivatives has been developed by way of a palladium-catalyzed decarboxylative cyclization of gamma-methylidene-delta-valerolactones with isatins. By employing a newly prepared phosphoramidite ligand, the reaction proceeds smoothly with excellent diastereoselectivity. Preliminary results of its application to asymmetric catalysis using a chiral phosphoramidite ligand are also described.
Sigma-Aldrich
δ-Valerolactone, technical grade