MilliporeSigma
Antioxidant effect of polyphenolic glabridin on LDL oxidation.
Toxicology and industrial health (2009-08-05)
E Carmeli, Y Fogelman
ABSTRACT
This study was conducted to determine the effect of a natural polyphenolic isoflavone antioxidant (Glabridin) on low-density lipoprotein (LDL) oxidation. Determination of the extent of LDL oxidation was done by measuring the formation of Thiobarbituric acid reactive substances (TBARS). After oral administration of licorice-root ethanol extract to healthy subjects for 6 months, the subjects' oxidative stress level as well as plasma LDL oxidation reduced by 20%. We concluded that dietary consumption of glabridin protects LDL from oxidation.
Sigma-Aldrich
Glabridin, ≥98% (HPLC), powder