MilliporeSigma
  • Iron-catalyzed selective oxidation of N-methyl amines: highly efficient synthesis of methylene-bridged bis-1,3-dicarbonyl compounds.
Iron-catalyzed selective oxidation of N-methyl amines: highly efficient synthesis of methylene-bridged bis-1,3-dicarbonyl compounds.
Organic letters (2009-08-18)
Haijun Li, Zhiheng He, Xingwei Guo, Wenjuan Li, Xuhui Zhao, Zhiping Li
ABSTRACT
Methylene-bridged bis-1,3-dicarbonyl derivatives were synthesized efficiently by iron-catalyzed oxidative reactions of 1,3-dicarbonyl compounds and N,N-dimethylaniline. Bipyrazoles and substituted 1,4-dihydropyridine were obtained by the reactions of bis-1,3-dicarbonyl compounds with hydrazines and ammonium acetate, respectively.
Sigma-Aldrich
N,N-Dimethylaniline, SAJ first grade, ≥99.0%
Sigma-Aldrich
N,N-Dimethylaniline, purified by redistillation, ≥99.5%
Sigma-Aldrich
N,N-Dimethylaniline, 99%
Sigma-Aldrich
N,N-Dimethylaniline, ReagentPlus®, 99%