MilliporeSigma
Synthesis of SnS quantum dots.
Journal of the American Chemical Society (2009-10-22)
Ying Xu, Najeh Al-Salim, Chris W Bumby, Richard D Tilley
ABSTRACT
Highly mondispersed SnS nanocrystals have been synthesized using ethanolamine ligands. SnS nanocrystals are small enough to be in the quantum confinement regime.