MilliporeSigma
  • Kinetics of the sonophotocatalytic degradation of orange G in presence of Fe(3+).
Kinetics of the sonophotocatalytic degradation of orange G in presence of Fe(3+).
Water science and technology : a journal of the International Association on Water Pollution Research (2009-10-22)
Jagannathan Madhavan, Franz Grieser, Muthupandian Ashokkumar
ABSTRACT
The degradation and mineralization of orange G (OG) in aqueous solution by means of ultrasound irradiation at a frequency of 213 kHz and its combination with Fe(3+) were investigated. The effect of Fe(3+) concentration on the degradation efficiency was studied. The degradation of the dye followed first-order like kinetics under the conditions examined. A comparison study on the hybrid technique of sonophotocatalytic degradation in the presence of Fe(3+) (SonoFenton) with the individual techniques of photocatalysis and sonolysis was also performed. A slight synergistic enhancement in the degradation of the dye was observed during the sonophotocatalytic oxidation of OG using Fe(3+). The total organic carbon (TOC) measurements, carried out in order to evaluate the mineralization efficiency of OG using sonolysis, photocatalysis and sonophotocatalysis, showed a synergetic effect of combining sonolysis and photolysis.
Sigma-Aldrich
Orange G, for NA electrophoresis
Sigma-Aldrich
Orange G, certified by the BSC
Sigma-Aldrich
Orange G, certified by the BSC, Dye content 80 %