MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Improved method for the synthesis of beta-carbonyl silyl-1,3-dithianes by the double conjugate addition of 1,3-dithiol to propargylic carbonyl compounds.
Improved method for the synthesis of beta-carbonyl silyl-1,3-dithianes by the double conjugate addition of 1,3-dithiol to propargylic carbonyl compounds.
The Journal of organic chemistry (2009-11-03)
Sumit Mukherjee, Dimitra Kontokosta, Aditi Patil, Sivakumar Rallapalli, Daesung Lee
ABSTRACT
Base-mediated double conjugate addition of 1,3-propane dithiol to various silylated propargylic aldehydes and ketones allows for an efficient and scalable synthesis of beta-carbonyl silyl-1,3-dithianes.
Sigma-Aldrich
Toluene-3,4-dithiol, technical grade, 90%
Supelco
Toluene-3,4-dithiol, for spectrophotometric det. of Mo, Sn, W, and also Ag and Re, ≥97.0%