MilliporeSigma
  • Gene clusters involved in isethionate degradation by terrestrial and marine bacteria.
Gene clusters involved in isethionate degradation by terrestrial and marine bacteria.
Applied and environmental microbiology (2009-11-26)
Sonja Weinitschke, Pia I Sharma, Ulrich Stingl, Alasdair M Cook, Theo H M Smits
ABSTRACT
Ubiquitous isethionate (2-hydroxyethanesulfonate) is dissimilated by diverse bacteria. Growth of Cupriavidus necator H16 with isethionate was observed, as was inducible membrane-bound isethionate dehydrogenase (IseJ) and inducible transcription of the genes predicted to encode IseJ and a transporter (IseU). Biodiversity in isethionate transport genes was observed and investigated by transcription experiments.