MilliporeSigma
[Study on the volatile oil of Murraya exotica].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2009-12-08)
Ping-Chuan Jiang, Jun Zhou, Bin Cao, Gui-Ning Wei, Kai-Jia He, Bu-Ming Liu
ABSTRACT
To study the volatile oil of Murraya exotica. The volatile oil of Murraya exotica was extracted by steam-stilling and was identified by GC-MS-DS. More than 90 compounds were separated, and 59 compounds were identified, accounting for 93.9% of the total essential oil of Murraya exotica. The major constituents of volatile oil were bicyclogermacrene (26.0%), beta-caryophyllene (20.8%), alpha-caryophyllene (5.8%), delta-cadinene (4.7%), spathulenol (4.3%), trans-alpha-bergamotene (4.1%), germacrene D (3.7%), beta-bisabolene (3.0%), ar-Curcumene (2.5%). The major components of Murraya exotica are the terpenoids, including 80.6% of sesquiterpenoids and 11.9% of monoterpenoids. Bicyclogermacrene is identified in Murraya genus for the first time.