MilliporeSigma
  • Purification of human IgG by negative chromatography on omega-aminohexyl-agarose.
Purification of human IgG by negative chromatography on omega-aminohexyl-agarose.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2010-01-19)
Maria Cristiane Martins de Souza, Igor Tadeu Lazzarotto Bresolin, Sonia Maria Alves Bueno
ABSTRACT
The omega-aminohexyl diamine immobilized as ligand on CNBr- and bisoxirane-activated agarose gel was evaluated for the purification of human immunoglobulin G (IgG) from serum and plasma by negative affinity chromatography. The effects of matrix activation, buffer system, and feedstream on recovery and purity of IgG were studied. A one-step purification process using Hepes buffer at pH 6.8 allowed a similar recovery (69-76%) of the loaded IgG in the nonretained fractions for both matrices, but the purity was higher for epoxy-activated gel (electrophoretically homogeneous protein with a 6.5-fold purification). The IgG and human serum albumin (HSA) adsorption equilibrium studies showed that the adsorption isotherms of IgG and HSA obeyed the Langmuir-Freundlich and Langmuir models, respectively. The binding capacity of HSA was high (210.4 mg mL(-1) of gel) and a positive cooperativity was observed for IgG binding. These results indicate that immobilizing omega-aminohexyl using bisoxirane as coupling agent is a useful strategy for rapid purification of IgG from human serum and plasma.
Sigma-Aldrich
ω-Aminohexyl–Agarose, saline suspension