MilliporeSigma
  • 2-(Phenylethyl)ammonium hydrogensquarate hemihydrate: crystal structure, solid-state IR-spectroscopic and theoretical characterization.
2-(Phenylethyl)ammonium hydrogensquarate hemihydrate: crystal structure, solid-state IR-spectroscopic and theoretical characterization.
Amino acids (2010-01-19)
Bojidarka B Ivanova, Rüdiger W Seidel, Tsonko Kolev, William Sheldrick, Michael Spiteller
ABSTRACT
The title compound, 2-(phenylethyl)ammonium hydrogensquarate hemihydrate, was synthesized and structurally and spectroscopically characterized by a single crystal X-ray diffraction and solid-state polarized IR spectroscopy of oriented colloids in a nematic host. The crystal structure consists of two crystallographically independent 2-(phenylethyl)ammonium cations, joined in a 2D hydrogen-bonded network with hydrogensquarate anions and solvent water molecules. Surprisingly, the crystallographically non-equivalent cations exhibit differing pseudo T and G trans configurations.
Sigma-Aldrich
3,4-Dihydroxy-3-cyclobutene-1,2-dione, ≥99.0% (HPLC)